Learn More
Chemglass Life Sciences Check Valve
SDP
Supplier:  Chemglass Life Sciences CG454001
Encompass_Preferred
Check Valve. Similar to CG-4539, but having serrated hose connections on either end to accept flexible tubing. Tubing O.D. is 10mm while the overall length is approx. 100mm. Hose connection O.D. is 9mm at the middle ring.
Catalog No. 50-153-1749