Learn More
Medchemexpress LLC Chelerythrine chloride | 3895-92-9 | 99.1% | 1 ML

Supplier: Medchemexpress LLC HY1204810MM 1ML
Chelerythrine chloride is a benzophenanthridine alkaloid and a cell-permeable inhibitor of protein kinase C used in biochemical and cell-based research. It is supplied as high-purity solid or as a ready-to-use 10 mM solution in DMSO.
- Cell-permeable protein kinase C inhibitor for biochemical and cellular assays.
- Available as solid powder and 10 mM solution in DMSO (1 mL).
- High purity suitable for research applications.
- Molecular formula C21H18ClNO4; molecular weight 383.82 g·mol⁻1.
- Intended for research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.