Learn More
Medchemexpress LLC Chelerythrine chloride | 3895-92-9 | 99.1% | 10 MG
SDP
Supplier:  Medchemexpress LLC HY1204810MG
Encompass_Preferred
Chelerythrine chloride is a benzophenanthridine alkaloid supplied as a high-purity research reagent and a potent, cell-permeable inhibitor of protein kinase C (PKC) used in biochemical and cellular studies.
  • High purity: 99.1% by HPLC.
  • Potent inhibitor of protein kinase C and reported to interact with Bcl-xL.
  • Supplied as a solid powder suitable for biochemical and cellular assays.
  • Soluble in DMSO, limited solubility in water.
  • Available in mg-scale packages, including 10 mg, for research use.
Catalog No. 50-002-31853