Learn More
Selleck Chemical LLC Chelerythrine-E0124-25MG
SDP
Supplier:  Selleck Chemical LLC E012425MG
Encompass_Preferred
Chelerythrine (Toddaline Broussonpapyrine) is a potent selective antagonist of PKC with an IC50 of 0 66 M Chelerythrine also inhibits the BclXL-Bak BH3 peptide binding with an IC50 of 1 5 M Chelerythrine shows antitumor antidiabetic and anti-inflammatory activity
Catalog No. 50-313-6652