Learn More
Chem-Impex International, Inc. CHEM IMPEX INTL INC Dimethyl Carbonate, 99.50% GC, 5 kg
Supplier:  Chem-Impex International, Inc. 369065KG
Encompass_Preferred
Dimethyl carbonate is a versatile and eco-friendly solvent and reagent widely utilized in various industrial applications. It is known for its low toxicity and excellent solvency properties, serving as a key component in the production of polycarbonate plastics. It is also employed as a methylating agent in organic synthesis for pharmaceuticals and agrochemicals, and acts as a green alternative to traditional toxic solvents.
  • Effective solvent and green alternative to toxic solvents.
  • Used in polycarbonate plastics production and as a fuel additive.
  • Versatile reagent in organic synthesis and for battery electrolytes.
Catalog No. NC3971926