Learn More
AVANTOR MASTERFLEX (VWR LLC) Masterflex I/P Precision Pump Tubing, Chem-Durance Bio, I/P 73; 50 ft
Supplier:  AVANTOR MASTERFLEX (VWR LLC) MFLX0644273/DEL
Encompass
Masterflex Chem-Durance Bio Precision pump tubing performs in a wide range of chemical applications, limiting the number of tubing formulations needed to keep on hand. The plasticizer-free, smooth-bore liner is hydrophobic and resists the absorption/adsorption of aqueous fluids. Ideal for laboratory, pharmaceutical processing, and biotech applications, this tubing complies with USP class VI and FDA standards. Tubing can be sterilized by autoclave, gamma radiation, or ethylene oxide.
Catalog No. NC0617018