Learn More
Medchemexpress LLC Chemerin-9 (149-157) (TFA) | 98.2% | 1177.18 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP1844A10MG
Encompass_Preferred
Chemerin-9 (149-157) TFA is a potent agonist of chemokine-like receptor 1 (CMKLR1) with anti-inflammatory activity. It stimulates phosphorylation of Akt and ERK, and promotes ROS production. This product also ameliorates Aβ1-42-induced memory impairment and regulates immune responses, adipocyte differentiation, and glucose metabolism.
  • Potent agonist of chemokine-like receptor 1 (CMKLR1)
  • Exhibits anti-inflammatory activity
  • Stimulates phosphorylation of Akt and ERK
  • Promotes ROS production
  • Ameliorates Aβ1-42-induced memory impairment
  • Regulates immune responses, adipocyte differentiation, and glucose metabolism
  • Stimulates migration in cardiac fibroblasts
  • Alleviates glucose intolerance and IR in PDM mice
  • Provides anti-inflammatory and anti-angiogenic effects in ApoE-/- mice
Catalog No. 50-004-23182