Learn More
Lakeland Industries Chemical Suits
Supplier:  Lakeland Industries C1T130Y5X
Encompass_Preferred
Lakeland's ChemMax 1 chemical suit with heat-sealed seams provides you with defense against light liquid splash in general industry, environmental cleaning, and chemical handling applications. This chemical coverall features an attached hood and storm flap. All ChemMax 1 products are constructed with a unique polyethylene barrier film that provides you with protection from harmful contaminants. Offered in cases of 6 each, C1T130Y is offered in sizes MD-4X.
Catalog No. 50-284-1246