Learn More
Cusabio Technology LLC chemokine C-C motif receptor 2
Supplier:  Cusabio Technology LLC CSBE14200H
Encompass_Preferred
This kit detects CCR2 in serum plasma tissue homogenates and cerebrospinal fluid (CSF) with a detection range of 23 44 pg/mL to 1500 pg/mL and a sensitivity of 5 86 pg/mL Utilizing a sandwich ELISA technique it provides quantitative results within 1 to 5 hours requiring only 50-100 L of sample per test The assay is measured at 450 nm ensuring reliable and precise detection of CCR2 levels in various biological samples Designed for researchers and clinicians this kit offers a robust tool for studying CCR2-related pathways and disease mechanisms supporting applications in immunology inflammation research and biomarker analysis Its standardized protocol ensures reproducibility across experimental and diagnostic settings
Catalog No. 50-000-51044