Learn More
Cusabio Technology LLC chemokine C-X-C motif ligand11
Supplier:  Cusabio Technology LLC CSBEL006241MO
Encompass_Preferred
This kit detects CXCL11 in mouse serum plasma cell culture supernates and tissue homogenates with a detection range of 31 25 pg/mL to 2000 pg/mL offering a sensitivity of 7 81 pg/mL Utilizing a quantitative sandwich ELISA methodology it requires a sample volume of 50-100 L and an incubation period of 1-5 hours The assay is measured at 450 nm providing reliable and precise results for research applications requiring quantification of CXCL11 levels in biological samples Its broad compatibility with multiple sample types makes it suitable for studies involving inflammatory responses immune regulation or related molecular pathways
Catalog No. 50-000-49957