Learn More
Medchemexpress LLC Chenodeoxycholic acid-13C | 52918-92-0 | 98.0% | C2313CH40O4 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY76847S2
Encompass_Preferred
Chenodeoxycholic acid-13C is a 13C-labeled version of Chenodeoxycholic Acid. It is a hydrophobic primary bile acid that activates nuclear receptors (FXR) involved in cholesterol metabolism.
  • Used as a tracer in various applications.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-58236