Learn More
Medchemexpress LLC Chenodeoxycholic acid 3-glucuronide | 58814-71-4 | 99.9% | 568.70 | C30H48O10 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW5858761MG
Encompass_Preferred
Chenodeoxycholic acid 3-glucuronide is a glucuronidated metabolite of chenodeoxycholic acid supplied as a research-grade biochemical. It acts as a farnesoid X receptor (FXR) modulator and is provided as a powder with documented purity, molecular information, and stability data for use in biochemical and cellular studies.
  • High purity (99.9%) suitable for research use.
  • Available in small mass quantities (1 mg, 5 mg, 10 mg) for assay development.
  • Documented molecular formula and molecular weight for analytical work.
  • COA, SDS, and data sheet provided for quality and safety assurance.
  • Recommended storage conditions support long-term stability.
  • Useful as an FXR-active metabolite in biochemical and cellular assays.
Catalog No. 50-002-84692