Learn More
Medchemexpress LLC Chenodeoxycholic acid 3-glucuronide | 58814-71-4 | 99.9% | 568.70 g/mol | C30H48O10 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYW58587610MG
Encompass_Preferred
Chenodeoxycholic acid 3-glucuronide is a metabolite of chenodeoxycholic acid supplied as a high-purity solid for laboratory research. It functions as an activator of the farnesoid X receptor (FXR) in biochemical and cell-based assays and is provided with storage and handling recommendations. For research use only; not for human or clinical use.
  • High purity solid powder: 99.9%.
  • Activates FXR; reported EC50 ≈8 μM in biochemical assays and ≈11 μM in HEK293T cells.
  • Available in small milligram pack sizes (1-100 mg).
  • Storage recommendations: powder -20°C for long term, 4°C for short term; in solvent store at -80°C for extended stability.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-002-84691