Learn More
Medchemexpress LLC Chenodeoxycholic Acid | 474-25-9 | 99.9% | C24H40O4 | 5 G
SDP
Supplier:  Medchemexpress LLC HY768475G
Encompass_Preferred
Chenodeoxycholic Acid is a hydrophobic primary bile acid that activates nuclear receptors (FXR) involved in cholesterol metabolism.
  • Activates nuclear receptors (FXR)
  • Inhibits 11 beta HSD2
  • Stimulates Ishikawa cell growth
  • Induces LDL receptor mRNA levels
  • Activates C/EBPβ
  • Enhances luciferase gene transcription
  • Activates AMP-activated protein kinase (AMPK)
Catalog No. 50-004-34039