Learn More
Medchemexpress LLC Chenodeoxycholic Acid | 474-25-9 | 99.9% | C24H40O4 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY76847500MG
Encompass_Preferred
Chenodeoxycholic Acid is a hydrophobic primary bile acid that activates nuclear receptors (FXR) involved in cholesterol metabolism. This product is intended for research use only.
  • Activates nuclear receptors (FXR) involved in cholesterol metabolism.
  • Inhibits 11 beta HSD2.
  • Stimulates Ishikawa cell growth by inducing Cyclin D1 protein and mRNA expression.
  • Induces LDL receptor mRNA levels approximately 4-fold.
  • Activates C/EBPβ and AMP-activated protein kinase (AMPK).
Catalog No. 50-004-34487