Learn More
Medchemexpress LLC Chenodeoxycholic Acid-d4 | 99102-69-9 | 99.9% | C24H36D4O4 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY76847S5MG
Encompass_Preferred
Chenodeoxycholic Acid-d4 is a deuterium-labeled primary bile acid. It is a hydrophobic compound that activates nuclear receptors (FXR) involved in cholesterol metabolism, and is intended for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-34489