Learn More
Medchemexpress LLC Chenodeoxycholic acid sodium | 2646-38-0 | 98.6% | C24H39NaO4 | 1 G
SDP
Supplier:  Medchemexpress LLC HY76847A1G
Encompass_Preferred
Chenodeoxycholic acid sodium is a hydrophobic primary bile acid that activates nuclear receptors (FXR) involved in cholesterol metabolism. It inhibits 11 beta HSD2 with an IC50 of 22 mM and causes cortisol-dependent nuclear translocation, increasing transcriptional activity of the mineralocorticoid receptor (MR). This compound is for research use only.
  • Inhibits 11 beta HSD2.
  • Increases MR transcriptional activity.
  • Stimulates Ishikawa cell growth via Cyclin D1 protein and mRNA expression.
  • Activates TGR5-dependent pathway.
  • Induces LDL receptor mRNA levels in Hep G2 cells.
  • Increases HMG-CoA reductase and synthase mRNA levels.
  • Activates C/EBPβ, enhancing luciferase gene transcription.
  • Activates AMPK, leading to ERK1/2 activation.
Catalog No. 50-004-34069