Learn More
Medchemexpress LLC Chenodeoxycholic acid sodium | 2646-38-0 | 98.6% | C24H39NaO4 | 5 G

Supplier: Medchemexpress LLC HY76847A5G
Chenodeoxycholic acid sodium is a hydrophobic primary bile acid that activates nuclear receptors (FXR) involved in cholesterol metabolism. This product is intended for research use only.
- Molecular weight: 414.55
- Appearance: Solid
- Color: White to off-white
- Structure classification: Steroids
- Initial source: Endogenous metabolite
- Solubility in DMSO: 100 mg/mL (241.23 mM); requires ultrasonic, use newly opened DMSO due to hygroscopic nature
- Solubility in H2O: 8.75 mg/mL (21.11 mM); requires ultrasonic and warming to 60°C
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.