Learn More
Medchemexpress LLC Chenodeoxycholic acid sodium | 2646-38-0 | 98.61% | C24H39NaO4 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY76847A100MG
Encompass_Preferred
Chenodeoxycholic acid sodium is a hydrophobic primary bile acid that activates nuclear receptors (FXR) involved in cholesterol metabolism. It is for research use only.
  • Activates nuclear receptors (FXR)
  • Inhibits 11 beta HSD2
  • Stimulates Ishikawa cell growth
  • Induces LDL receptor mRNA levels
  • Activates C/EBPβ
  • Activates AMP-activated protein kinase (AMPK)
Catalog No. 50-004-33769