Learn More
Medchemexpress LLC Chenodeoxycholic acid sodium | 2646-38-0 | MFCD00065452 | 98.6% | C24H39NaO4 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY76847A10MM/1ML
Encompass_Preferred
Chenodeoxycholic acid sodium (CDCA sodium) is a hydrophobic primary bile acid intended for research use. It activates nuclear receptors (FXR) involved in cholesterol metabolism. In vitro studies demonstrate its ability to inhibit 11β-HSD2, stimulate cell growth by inducing Cyclin D1 expression via the TGR5-dependent pathway, and increase LDL receptor and HMG-CoA reductase mRNA levels. It also induces short-circuit current (Isc) and activates C/EBPβ and AMP-activated protein kinase (AMPK).
  • Activates nuclear receptors (FXR) for cholesterol metabolism.
  • Inhibits 11β-HSD2 activity.
  • Stimulates Ishikawa cell growth.
  • Increases LDL receptor and HMG-CoA reductase mRNA levels.
  • Activates C/EBPβ and AMP-activated protein kinase (AMPK).
Catalog No. 50-004-34488