Learn More
Cambridge Isotope Laboratories CHENODEOXYCHOLIC ACID UN-50MG
Supplier:  Cambridge Isotope Laboratories ULM95400.05
Encompass
CHENODEOXYCHOLIC ACID UNLABELED, 50MG, CAS# 474-25-9, MOLWT: 392.57
Catalog No. NC2001605