Learn More
Cayman Chemical ChenodeoxycholIc AcIdodIum 5g
SDP
Supplier:  Cayman Chemical 353465 G
Encompass_Preferred
A primary bile acid; an FXR agonist that binds to FXRs in a TR-FRET assay (EC50 = 13 µM) and induces FXR transactivation in a reporter assay; induces transcription of GCL in primary hepatocytes and HepG2 cells from 25-100 µM
Catalog No. 50-291-3398