Learn More
OriGene CHI3L1 CytoSection 25 Slides
SDP
Supplier:  OriGene TS403769P5
Encompass_Preferred
Transient overexpression of CHI3L1 in HEK293T cells, FFPE control for IHC, ICC and ISH staining, 25 slides per pack
Catalog No. NC2944463