Learn More
Kingfisher Biotech Inc Multi-species (Bovine, Caprine, Ovine) IL-5 ELISpot - Up to two (2) ELISpot assays
SDP
Supplier:  Kingfisher Biotech Inc SPOT2392C004
Encompass_Preferred
Multi-species (Bovine, Caprine, Ovine) IL-5 ELISpot ELISpot contains Capture Antibody, Standard Control, and Detection Antibody for development of up to two (2) ELISpot assays. Other required materials and reagents are not supplied. The antibodies have been determined to function in an ELISpot with the Standard Control provided. Components may also be purchased separately. For research use only. Made in the USA
Catalog No. 50-289-2000