Learn More
Immunology Consultants Chicken IgY ELISA Kit
SDP
Supplier:  Immunology Consultants E30Y
Encompass_Preferred
The Chicken IgY test kit is a highly sensitive two-site enzyme linked immunoassay (ELISA) for measuring IgY in chicken biological samples
Catalog No. 50-240-6565