Learn More
American Research Products Inc Chicken Immunoglobulin A IgA
Supplier:  American Research Products Inc CSBE11232CH
Encompass
Chicken Immunoglobulin A IgA ELISA Kit from CUSABIO Detection Range 781 25 ng/mL-50000 ng/mL Sensitivity 390 ng/mL Sample serum plasma Method Competitive
Catalog No. 50-003-29079