Learn More
Cusabio Technology LLC CHICKEN interleukin 10 ELISA
Supplier:  Cusabio Technology LLC CSBE12835C
Encompass_Preferred
This kit detects IL10 in chicken serum plasma and tissue homogenates using a competitive ELISA method providing quantitative results with a detection range of 1 pg/mL to 200 pg/mL and a sensitivity of 0 5 pg/mL The assay requires 50-100 L of sample per test and can be completed within 1-5 hours Measurements are performed at an absorbance wavelength of 450 nm ensuring reliable detection of IL10 levels in various biological matrices Designed for research applications this kit offers a standardized protocol to accurately quantify IL10 concentrations supporting studies on immune response and cytokine profiling in avian models
Catalog No. 50-000-48845