Learn More
Biomatik Corporation Chicken N-Terminal Pro-Brain Natriuretic Peptide (NT-ProBNP) ELISA Kit
Supplier:  Biomatik Corporation EKU11317
Encompass_Preferred
Chicken N-Terminal Pro-Brain Natriuretic Peptide (NT-ProBNP) ELISA Kit for detecting NT-ProBNP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Chicken (Gallus). Detection Range: 123.5-10,000pg/ml. Sensitivity: 44.3pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P18908. Gene ID: 395765. Target Synonyms: NT-Pro-BNP; ; N-BNP. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8168