Learn More
Sino Biological Chikungunya virus (CHIKV) (UVE/CHIKV/2024/RE/CNR_79903) E2 / Envelope 2 ELISA Pair Set 5 Plates
Supplier:  Sino Biological SEK410945
Encompass_Preferred
The Chikungunya virus (CHIKV) (UVE/CHIKV/2024/RE/CNR_79903) E2 / Envelope 2 ELISA Pair Set is for the quantitative determination of Chikungunya virus (CHIKV) (UVE/CHIKV/2024/RE/CNR_79903) E2 / Envelope 2.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Chikungunya virus (CHIKV) (UVE/CHIKV/2024/RE/CNR_79903) E2 / Envelope 2 coated on a 96-well plate. Standards and samples are added to the wells, and any Chikungunya virus (CHIKV) (UVE/CHIKV/2024/RE/CNR_79903) E2 / Envelope 2 present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated mouse anti-Chikungunya virus (CHIKV) (UVE/CHIKV/2024/RE/CNR_79903) E2 / Envelope 2 monoclonal antibody is then added, producing an antibody-antigen-antibody ""sandwich"". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Chikungunya virus (CHIKV) (
Catalog No. 50-007-85859