Learn More
Cayman Chemical ChIkusetsusaponIn IVa 50mg
SDP
Supplier:  Cayman Chemical 3483250 MG
Encompass_Preferred
An oleanane-type triterpenoid with diverse biological activities; inhibits the growth of HL-60 and HCT116 cells (IC50s = 76.23 and 78.11 µM, respectively); reduces LPS-induced increases in iNOS, COX-2, IL-1β, IL-6, and TNF-α levels in PMA-differentiated THP-1 macrophages at 50-200 UG/ml; decreases plasma glucose, triglyceride, and LDL-cholesterol, and increases HDL-cholesterol, levels in a rat model of STZ- and high-fat diet-induced type 2 diabetes at 45-180 mg/kg
Catalog No. 50-291-1044