Learn More
Grainger Child Toilet Bowl Round Floor FlushValve
Supplier:  Grainger 16U007
Encompass_Preferred
The American Standard child toilet bowl is a vitreous china toilet bowl designed for children ages 3-10. It features an elongated bowl shape with a standard height of 14.5 inches. The rough-in/outlet dimension is 12 inches. This toilet bowl has a 2-1/8 inch fully-glazed trapway and is designed for use with the American Standard Cadet 3 Right Height tank. It meets EPA WaterSense criteria to use 20% less water than standard 1.6 gpf toilets. This white toilet bowl comes with a 5 year limited warranty.
Catalog No. 50-276-5329