Learn More
Enzo Life Sciences CHIP polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ENZABS2730100
Encompass_Preferred
Isotype: IgG. Immunogen: Synthetic peptide corresponding to aa 218-232 of human CHIP. Species reactivity: Human, Mouse. Applications: WB. Purity: Antigen affinity chromatography. Formulation: Liquid. In PBS containing 1mg/ml BSA and 0.05% sodium azide.
Catalog No. 50-201-2817