Learn More
Apexbio Technology LLC CHIR-090, 728865-23-4, MFCD22665727, 10 mg
SDP
Supplier:  Apexbio Technology LLC A330710
Encompass_Preferred
MF: C24H27N3O5, MW: 437.49, Solubility: 21.85 mg/mL in DMSO,insoluble in EtOH,insoluble in H2O. CHIR-090 is a very potent, low, tight-binding inhibitor of LpxC with Ki value of 4.0 nM.
Catalog No. 50-115-0551