Learn More
BIOGEMS INTERNATIONAL INC CHIR 98014 5mg
SDP
Supplier:  BIOGEMS INTERNATIONAL INC 55639935MG
Encompass
CHIR 98014 is a potent and reversible inhibitor of Glycogen Synthase Kinase 3 (GSK3), inhibiting both the alpha (IC50: 0.65 nM) and beta (IC50: 0.58 nM) isoforms. GSK3 functions by phosphorylating a serine or threonine residue on its target substrate and is part of the Wnt pathway that signals the cell to divide and proliferate. CHIR 98014 activates glycogen synthase in cells to improve glucose disposal and to reduce tau phosphorylation in neurons. It is reported to stimulate vascular cell differentiation of hPSC and enhances the transformation of vascular smooth muscle cells into osteogenic-like cells under high phosphate conditions.
Catalog No. 50-153-0186