Learn More
TARGETMOL CHEMICALS INC CHIR-99021 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T231025MG
Encompass_Preferred
Also available in 1 mL, 2 mg, 5 mg, 10 mg, 50 mg, 100 mg, 200 mg, 500 mg and bulk. Please contact Fisher for quotes. CHIR-99021 (CT99021) is an activator of the Wnt/beta-catenin signaling pathway and a GSK-3alpha/beta inhibitor (IC50=10/6.7 nM) with selective and oral activity.CHIR-99021 induces cellular autophagy, which enhances self-renewal in mouse and human embryonic stem cells. Purity 99.29%
Catalog No. 50-287-7722