Learn More
Apexbio Technology LLC CHIR-99021 (CT99021), 100mg. Cas: 252917-06-9 MFCD: MFCD11846251. GSK-3 inhibitor, Cell-permeable, ATP-competitive
SDP
Supplier:  Apexbio Technology LLC A3011100
Encompass_Preferred
CHIR-99021 is a highly specific glycogen synthase kinase-3 (GSK-3) inhibitor which can inhibit both isoforms with IC50 of 10 nM (GSK-3) and 6.7 nM (GSK-3). Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-4779