Learn More
Sigma Aldrich Fine Chemicals Biosciences CHIR99021, 252917-06-9, MFCD11846251, 25 mg
Supplier:  Sigma Aldrich Fine Chemicals Biosciences SML104625MG
Encompass
Empirical Formula (Hill Notation): C22H18Cl2N8, Molecular Weight: 465.34, ≥98% (HPLC), storage temp: −20°C, Synonym: 6-[[2-[[4-(2,4-Dichlorophenyl)-5-(5-methyl-1H-imidazol-2-yl)-2-pyrimidinyl]amino]ethyl]amino]-3-pyridinecarbonitrile, CT99021.
Catalog No. NC0940125