Learn More
Cayman Chemical CHIR99021hydrochlorIde 10mg
SDP
Supplier:  Cayman Chemical 3917610 MG
Encompass_Preferred
An aminopyrimidine derivative that inhibits GSK3α and GSK3β (IC50s = 10 and 6.7 nM, respectively); activates glycogen synthesis in CHO-IR cells (EC50 = 0.763 μM) and in isolated type 1 diabetic rat skeletal muscle; has been shown to promote self-renewal of embryonic stem cells
Catalog No. 50-291-8355