Learn More
Chiral Technologies, Inc. CHIRALCEL OJ-3R 150X4.6 3UM
Supplier:  Chiral Technologies, Inc. 17824
Encompass
Coated Polysaccharid CHIRALCEL OJ-3R, 3 micron, 4.6 mm ID x 150 mm LN HPLC Reverse phase analytical column
Catalog No. NC0312457