Learn More
Chiral Technologies, Inc. CHIRALPAK AD-H SFC 4.6X100MM
Supplier:  Chiral Technologies, Inc. 19423
Encompass
Coated Polysaccharide, CHIRALPAK AD-H, 5 micron,4.6 mm ID x 100mm LN, SFC analytical column
Catalog No. NC0166704