Learn More
Chiral Technologies, Inc. CHIRALPAK AS-3 4.6X150MM 3 MIC
Supplier:  Chiral Technologies, Inc. 20524
Encompass
Coated Polysaccharid CHIRALPAK AS-3, 3 micron, 4.6 mm ID x 150 mm LN HPLC analytical column
Catalog No. NC0243762