Learn More
Chiral Technologies, Inc. CHIRALPAK AY-H (250X4.6MM
Supplier:  Chiral Technologies, Inc. 47325
Encompass
Coated Polysaccharide, CHIRALPAK AY-H, 5 micron,4.6 mm ID x 250mm LN, analytical column
Catalog No. NC9869662