Learn More
Chiral Technologies, Inc. CHIRALPAK IC COLUMN 4.6X150MM
Supplier:  Chiral Technologies, Inc. 83324
Encompass
Immobilized Polysaccharide, CHIRALPAK IC, 5 micron, 4.6 mm ID x 150 mm LN Analytical column
Catalog No. NC9566534