Learn More
Chiral Technologies, Inc. CHIRALPAK ZWIX(+) 4X250
Supplier:  Chiral Technologies, Inc. 51515
Encompass
Immobilized Zwitterionic Columns, CHIRALPAK ZWIX(+), 3 micron, 4.0mm ID x 250mm LN, Analytical column
Catalog No. 50-713-412