Learn More
Chiral Technologies, Inc. Chiralpak IE-3, High-Performance HPLC Column, Universal Solvent Compatibility, Maximum Pressure 300 bar
Supplier:  Chiral Technologies, Inc. 85523
Encompass
The CHIRALPAK IE-3 is a high-performance HPLC column designed for efficient chiral separations. Its unique immobilized amylose-based stationary phase provides robust performance and universal compatibility with solvents in various chromatography modes, including normal phase, reversed phase, and SFC. With exceptional stability against 'forbidden' solvents and a maximum temperature of 40 °C, this column supports advanced method development, ensuring reliable and reproducible results for analytical applications.
Catalog No. NC0532158