Learn More
Medchemexpress LLC Chitin, from crab carapace | 1398-61-4 | 50 G
SDP
Supplier:  Medchemexpress LLC HY12638950G
Encompass_Preferred
Chitin, from crab carapace is a long-chain polymer of N-acetylglucosamine with β-(1-4) linkages found in the exoskeleton of crabs. It inhibits the activation of NF-κB p65, alters its translocation to the nucleus, and interacts with the cell wall of *Candida* species. It exerts antifungal and anti-inflammatory effects and can be used in the research of gastric ulcer and candidiasis.
  • Inhibits NF-κB p65 activation and nuclear translocation.
  • Interacts with *Candida* species cell walls.
  • Exhibits antifungal activity against clinical isolates of *Candida* spp.
  • Provides anti-inflammatory effects.
  • Reduces gastric ulcer index in animal models.
  • Useful for research on gastric ulcer and candidiasis.
Catalog No. 50-004-38024