Learn More
Sigma Aldrich Fine Chemicals Biosciences Chitinase from Streptomyce
Supplier:  Sigma Aldrich Fine Chemicals Biosciences SAE015810UN
Encompass
Chitinase is an extracellular complex of enzymes that degrade chitin. Chitin is a cell wall component of fungi and exoskeletal essentials of different organisms which reshape their own chitin or digest/dissolve the chitin of other organisms (insects fungi yeast and algae and in the internal structures of other vertebrates). Chitinases have been detected in many microorganisms and in plants. In fungi chitinases assist in morphogenesis to break down the inherent chitin content of fungal cell walls. Plant chitinases help in resistance to fungal attack and counteracting fungal growth by targeting those same fungal cell walls. In bacteria bacterial chitinases assist in utilizing chitin as a carbon source and as an energy source. Streptomyces griseus produces multiple chitinases of different molecular masses after growth induction with chitin as the carbon source. The enzymatic hydrolysis of chitin to N-acetyl-D-glucosamine involves two consecutive enzyme reactions bullnbspThe first
Catalog No. 50-290-1163