Learn More
Medchemexpress LLC Chitosan (≥80% deacetylated, High viscosity,>400mPa.s) | 9012-76-4 | ≥80% | 50 G
SDP
Supplier:  Medchemexpress LLC HYB2144F50G
Encompass_Preferred
Chitosan (Deacetylated chitin; Poly(D-glucosamine)) (≥80% deacetylated, High viscosity,>400mPa.s) is a polysaccharide obtained by deacetylating chitin. It is a polycationic linear polysaccharide and a versatile biomaterial due to its non-toxicity, low allergenicity, biocompatibility, and biodegradability. This product exhibits antimicrobial activity against various bacteria and fungi.
  • Exhibits antimicrobial activity against various bacteria and fungi.
  • Possesses antitumor, antibacterial, antifungal, and antioxidant activities.
  • Versatile biomaterial known for its non-toxicity.
  • Low allergenicity, biocompatibility, and biodegradability.
Catalog No. 50-004-10922