Learn More
Medchemexpress LLC Chitosan (85%~90% deacetylated, mushroom) | 9012-76-4 | 85%-90% deacetylated | 10 G
SDP
Supplier:  Medchemexpress LLC HYB2144K10G
Encompass_Preferred
Chitosan (85%-90% deacetylated, mushroom) is a polysaccharide obtained by deacetylating chitin from mushroom. It is a polycationic linear polysaccharide derived from chitin, known as a versatile biomaterial.
  • Obtained by deacetylating chitin from mushroom
  • Polycationic linear polysaccharide derived from chitin
  • Exhibits non-toxicity, low allergenicity, biocompatibility, and biodegradability
  • Possesses antitumor, antibacterial, antifungal, and antioxidant activities
Catalog No. 50-004-12270