Learn More
Medchemexpress LLC Chitosan (≥85% deacetylated, viscosity>90 mPa.s, MW 15000) | 9012-76-4 | ≥85% deacetylated | 5 G
SDP
Supplier:  Medchemexpress LLC HYB2144K5G
Encompass_Preferred
Chitosan is a polysaccharide derived from deacetylating chitin. It demonstrates antimicrobial activity against various bacteria and fungi. This polycationic linear polysaccharide is a versatile biomaterial due to its non-toxicity, low allergenicity, biocompatibility, and biodegradability. Chitosan also possesses antitumor, antibacterial, antifungal, and antioxidant activities.
  • Polysaccharide obtained by deacetylating chitin
  • Exhibits antimicrobial activity against various bacteria and fungi
  • Versatile biomaterial
  • Non-toxicity
  • Low allergenicity
  • Biocompatibility
  • Biodegradability
  • Antitumor activity
  • Antibacterial activity
  • Antifungal activity
  • Antioxidant activity
Catalog No. 50-004-12375